2-Mercapto-5-methoxybenzimidazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methoxy-2-benzimidazolethiol, 1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione, 5-Methoxy-1H-benzo[d]imidazole-2-thiol, 5-Methoxybenzimidazole-2-thiol, 2-Mercapto-5-methoxy-1H-benzimidazole
OR9581-100G OR9581-25G OR9581-500G
APOSOR9581-100GEA 24.8 EUR
APOSOR9581-100G APOSOR9581-25G APOSOR9581-500G
2-Mercapto-5-methoxybenzimidazole 98%
2-Mercapto-5-methoxybenzimidazole
Formula: C₈H₈N₂OS
Melting Pt: 255…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00134581
CAS Number: 37052-78-1
EINECS: 253-326-2