4-Amino-3-ethylbenzonitrile
Supplier: Apollo Scientific
Warning
OR9583-1G OR9583-5G
APOSOR9583-1GEA 34.8 EUR
APOSOR9583-1G APOSOR9583-5G
4-Amino-3-ethylbenzonitrile
4-Amino-3-ethylbenzonitrile
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Melting Pt: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00041431
CAS Number: 170230-87-2
UN: 3439
ADR: 6.1 III