(1-Ethyl-1H-pyrazol-4-yl)methanol 95%
Supplier: Apollo Scientific
OR960077-1G OR960077-250MG OR960077-5G
APOSOR960077-1GEA 173 EUR
APOSOR960077-1G APOSOR960077-250MG APOSOR960077-5G
(1-Ethyl-1H-pyrazol-4-yl)methanol 95%
(1-Ethyl-1H-pyrazol-4-yl)methanol
Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02253733
CAS Number: 905307-04-2