6-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine 98%

Supplier: Apollo Scientific

OR960188-1G OR960188-5G
APOSOR960188-1GEA 219 EUR
APOSOR960188-1G APOSOR960188-5G
6-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine 98%
6-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine
Formula: C₉H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD11603432
CAS Number: 1082837-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR