[3-(2-Methylphenyl)-1,2,4-oxadiazol-5-yl]methylamine hydrochloride 96%
Supplier: Apollo Scientific
Synonyms: [3-(2-Methylphenyl)-1,2,4-oxadiazol-5-yl]methanamine
OR960237-1G OR960237-5G
APOSOR960237-1GEA 183 EUR
APOSOR960237-1G APOSOR960237-5G
[3-(2-Methylphenyl)-1,2,4-oxadiazol-5-yl]methylamine hydrochloride 96%
[3-(2-Methylphenyl)-1,2,4-oxadiazol-5-yl]methylamine hydrochloride
Formula: C₁₀H₁₂ClN₃O
MW: 225.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12027097
CAS Number: 937665-65-1