3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 96%
Supplier: Apollo Scientific
Synonyms: 3-Hydroxycyclohexanecarboxylic acid, 3-Hydroxycyclohexane-1-carboxylic acid
OR960353-1G OR960353-5G
APOSOR960353-1GEA 169 EUR
APOSOR960353-1G APOSOR960353-5G
3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 96%
3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19228828
CAS Number: 606488-94-2