1-(1,3-Dimethyl-1H-pyrazol-5-yl)ethanone 95%
Supplier: Apollo Scientific
OR960698-1G OR960698-250MG OR960698-5G
APOSOR960698-1GEA 348 EUR
APOSOR960698-1G APOSOR960698-250MG APOSOR960698-5G
1-(1,3-Dimethyl-1H-pyrazol-5-yl)ethanone 95%
1-(1,3-Dimethyl-1H-pyrazol-5-yl)ethanone
Formula: C₇H₁₀N₂O
Storage Temperature: Ambient
CAS Number: 87375-38-0