6,7-Dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione hydrochloride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1,5,6,7-Tetrahydropyrrolo[3,4-d]pyrimidine-2,4-dione hydrochloride
OR960748-100MG OR960748-1G OR960748-250MG
APOSOR960748-100MGEA 134 EUR
APOSOR960748-100MG APOSOR960748-1G APOSOR960748-250MG
6,7-Dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione hydrochloride 95%
6,7-Dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione hydrochloride
Formula: C₆H₈ClN₃O₂
MW: 189.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11616742
CAS Number: 13931-24-3