3,5-Dibromo-1H-1,2,4-triazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dibromo-1,2,4-triazole
OR9608-25G OR9608-5G
APOSOR9608-25GEA 46.4 EUR
APOSOR9608-25G APOSOR9608-5G
3,5-Dibromo-1H-1,2,4-triazole 98%
3,5-Dibromo-1H-1,2,4-triazole
Formula: C₂HBr₂N₃
MW: 226.8600 g/mol
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00487260
CAS Number: 7411-23-6
UN: 2811
ADR: 6.1,III