Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid 95%
Supplier: Apollo Scientific
Synonyms: 8-hydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid, 2-pyrazolo[1,5-a]pyrimidinecarboxylic acid, Pyrazolo[1,5-a]pyrimidine-2-carboxylicacid
OR961056-1G OR961056-250MG OR961056-5G
APOSOR961056-1GEA 78.3 EUR
APOSOR961056-1G APOSOR961056-250MG APOSOR961056-5G
Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid 95%
Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Formula: C₇H₅N₃O₂
MW: 163.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00297221
CAS Number: 378211-85-9