3-Phenyl-2-(phenylsulphonyl)-1,2-oxaziridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Benzenesulphonyl)-3-phenyloxaziridine
OR961468-1G OR961468-5G OR961468-10G
APOSOR961468-1GEA 132 EUR
APOSOR961468-1G APOSOR961468-5G APOSOR961468-10G
3-Phenyl-2-(phenylsulphonyl)-1,2-oxaziridine 95%
3-Phenyl-2-(phenylsulphonyl)-1,2-oxaziridine
Formula: C13H11NO3S
Storage Temperature: Freezer
CAS Number: 63160-13-4