1-Chloro-3,3-dimethylbutanone 95%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Chloropinacolone
OR961856-25ML OR961856-500ML
APOSOR961856-25MLEA 79.9 EUR
APOSOR961856-25ML APOSOR961856-500ML
1-Chloro-3,3-dimethylbutanone 95%
1-Chloro-3,3-dimethylbutanone
Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 170…173 °C
Melting Pt: –1 °C
Density: 1.03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035688
CAS Number: 13547-70-1
UN: 2810
ADR: 6.1 II