5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine, 2,5-Dibromo-3,4-ethylenedioxythiophene
OR9626-1G OR9626-250MG
APOSOR9626-1GEA 29 EUR
APOSOR9626-1G APOSOR9626-250MG
5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine 97%
5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine
Formula: C₆H₄Br₂O₂S
MW: 299.9700 g/mol
Melting Pt: 96…97 °C
Storage Temperature: Ambient
MDL Number: MFCD01651770
CAS Number: 174508-31-7