2-(2-Methoxyethoxy)benzoic acid 95%
Supplier: Apollo Scientific
OR962706-1G OR962706-250MG OR962706-5G
APOSOR962706-1GEA 255 EUR
APOSOR962706-1G APOSOR962706-250MG APOSOR962706-5G
2-(2-Methoxyethoxy)benzoic acid 95%
2-(2-Methoxyethoxy)benzoic acid
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08700372
CAS Number: 98684-32-3