2-Chlorothieno[3,2-d]pyrimidine 98%
Supplier: Apollo Scientific
OR963899-1G OR963899-250MG
APOSOR963899-1GEA 132 EUR
APOSOR963899-1G APOSOR963899-250MG
2-Chlorothieno[3,2-d]pyrimidine 98%
2-Chlorothieno[3,2-d]pyrimidine
Formula: C₆H₃ClN₂S
MW: 170.6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12406127
CAS Number: 1119280-68-0