2-Furan-2-ylethylamine 95%
Supplier: Apollo Scientific
OR963942-1G OR963942-250MG OR963942-5G
APOSOR963942-1GEA 219 EUR
APOSOR963942-1G APOSOR963942-250MG APOSOR963942-5G
2-Furan-2-ylethylamine 95%
2-Furan-2-ylethylamine
Formula: C₆H₉NO
MW: 111.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04113973
CAS Number: 1121-46-6