3,3-Dimethylindolin-2-one 97%
Supplier: Apollo Scientific
OR964406-1G OR964406-5G
APOSOR964406-1GEA 58.7 EUR
APOSOR964406-1G APOSOR964406-5G
3,3-Dimethylindolin-2-one 97%
3,3-Dimethylindolin-2-one
Formula: C10H11NO
Storage Temperature: Ambient
MDL Number: MFCD00158549
CAS Number: 19155-24-9