Boc-D-Glu(Obzl)-OH
Supplier: Apollo Scientific
Warning
Synonyms: Boc-D-Glu(OBzl)-OH, N-Boc-D-glutamic acid 5-benzyl ester
OR964505-100G OR964505-25G OR964505-5G
APOSOR964505-100GEA 459 EUR
APOSOR964505-100G APOSOR964505-25G APOSOR964505-5G
Boc-D-Glu(Obzl)-OH
Boc-D-Glu(Obzl)-OH
Formula: C₁₇H₂₃NO₆
MW: 337.372 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038258
CAS Number: 35793-73-8