2-Isopropoxyaniline 97%
Supplier: Apollo Scientific
OR964694-1G OR964694-25G
APOSOR964694-1GEA 29 EUR
APOSOR964694-1G APOSOR964694-25G
2-Isopropoxyaniline 97%
2-Isopropoxyaniline
Formula: C₉H₁₃NO
MW: 151.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00242947
CAS Number: 29026-74-2