3-Butyn-1-amine hydrochloride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Butynylamine hydrochloride, 3-Butyn-1-ylammonium chloride, But-3-yn-1-ylammonium chloride, But-3-yn-1-amine hydrochloride
OR964735-250MG OR964735-5G OR964735-25G
APOSOR964735-250MGEA 0 EUR
APOSOR964735-250MG APOSOR964735-5G APOSOR964735-25G
3-Butyn-1-amine hydrochloride 95%
3-Butyn-1-amine hydrochloride
Formula: C₄H₇N·HCl
MW: 105.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658391
CAS Number: 88211-50-1