5,6-Dimethoxynicotinaldehyde 98%
Supplier: Apollo Scientific
OR964832-1G OR964832-250MG
APOSOR964832-1GEA 719 EUR
APOSOR964832-1G APOSOR964832-250MG
5,6-Dimethoxynicotinaldehyde 98%
5,6-Dimethoxynicotinaldehyde
Formula: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD11857649
CAS Number: 52605-99-9