2-(3-Hydroxyphenyl)ethanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxyphenethyl alcohol
OR965089-1G OR965089-5G
APOSOR965089-1GEA 66.7 EUR
APOSOR965089-1G APOSOR965089-5G
2-(3-Hydroxyphenyl)ethanol 95%
2-(3-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 168…173 °C
Density: 1.082 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 13398-94-2