2-Bromo-6-methoxyimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
OR965427-250MG OR965427-1G
APOSOR965427-250MGEA 437 EUR
APOSOR965427-250MG APOSOR965427-1G
2-Bromo-6-methoxyimidazo[1,2-a]pyridine
2-Bromo-6-methoxyimidazo[1,2-a]pyridine
Formula: C₈H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD12922819
CAS Number: 1042141-33-2