4-Morpholin-4-ylmethylthiazol-2-ylamine 97%
Supplier: Apollo Scientific
Synonyms: 4-(Morpholinomethyl)thiazol-2-amine
OR965574-100MG OR965574-1G OR965574-250MG
APOSOR965574-100MGEA 69.6 EUR
APOSOR965574-100MG APOSOR965574-1G APOSOR965574-250MG
4-Morpholin-4-ylmethylthiazol-2-ylamine 97%
4-Morpholin-4-ylmethylthiazol-2-ylamine
Formula: C₈H₁₃N₃OS
MW: 199.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06340096
CAS Number: 3008-61-5