5-Methylisoxazole-3-carboxaldehyde 97%
Supplier: Apollo Scientific
Danger
OR9680-1G OR9680-5G
APOSOR9680-1GEA 125 EUR
APOSOR9680-1G APOSOR9680-5G
5-Methylisoxazole-3-carboxaldehyde 97%
5-Methylisoxazole-3-carboxaldehyde
Formula: C₅H₅NO₂
MW: 111.1 g/mol
Boiling Pt: 53…57 °C/8mmHg
Melting Pt: 30…34 °C
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD06797459
CAS Number: 62254-74-4
UN: 1325
ADR: 4.1,III