2,4-Dichloroaniline 99%
Supplier: Apollo Scientific
Danger
OR9687-1KG OR9687-250G OR9687-100G
APOSOR9687-1KGEA 393 EUR
APOSOR9687-1KG APOSOR9687-250G APOSOR9687-100G
2,4-Dichloroaniline 99%
2,4-Dichloroaniline
Formula: C₆H₅Cl₂N
Boiling Pt: 245 °C
Melting Pt: 60…62 °C
Density: 1.567 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS Number: 554-00-7
EINECS: 209-057-8
UN: 3442
ADR: 6.1,II