3-Bromo-1-benzofuran 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromobenzofuran
OR9692-250MG
APOSOR9692-250MGEA 21.9 EUR
APOSOR9692-250MG
3-Bromo-1-benzofuran 97%
3-Bromo-1-benzofuran
Formula: C₈H₅BrO
MW: 197.04 g/mol
Boiling Pt: 110 °C
Melting Pt: 35…40 °C
Storage Temperature: Freezer
MDL Number: MFCD01029425
CAS Number: 59214-70-9