3-Chlorothioanisole 97%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Chlorophenyl)(methyl)sulphane, 3-Chlorophenyl methyl sulphide, 1-Chloro-3-(methylthio)benzene
OR9711-100G OR9711-25G OR9711-5G
APOSOR9711-100GEA 364 EUR
APOSOR9711-100G APOSOR9711-25G APOSOR9711-5G
3-Chlorothioanisole 97%
3-Chlorothioanisole
Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 218 °C
Density: 1.206 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040127
CAS Number: 4867-37-2