3-Bromo-2-nitrophenol 96%
Supplier: Apollo Scientific
Warning
OR9737-1G OR9737-5G
APOSOR9737-1GEA 34.8 EUR
APOSOR9737-1G APOSOR9737-5G
3-Bromo-2-nitrophenol 96%
3-Bromo-2-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.0100 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD08445675
CAS Number: 76361-99-4