1,3-Dichloro-2-nitrobenzene
Supplier: Apollo Scientific
Warning
OR9743-100G OR9743-250G OR9743-25G
APOSOR9743-100GEA 114 EUR
APOSOR9743-100G APOSOR9743-250G APOSOR9743-25G
1,3-Dichloro-2-nitrobenzene
1,3-Dichloro-2-nitrobenzene
Formula: C₆H₃Cl₂NO₂
Boiling Pt: 130 °C/8mmHg
Melting Pt: 65…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00456795
CAS Number: 601-88-7
UN: 2811
ADR: 6.1,III