2-(4-Chloro-2-methylphenoxy)acetohydrazide
Supplier: Apollo Scientific
Warning
OR9774-25G OR9774-5G
APOSOR9774-25GEA 161 EUR
APOSOR9774-25G APOSOR9774-5G
2-(4-Chloro-2-methylphenoxy)acetohydrazide
2-(4-Chloro-2-methylphenoxy)acetohydrazide
Formula: C₉H₁₁ClN₂O₂
MW: 214.65 g/mol
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00082720
CAS Number: 32022-38-1