2-Chloro-6-methylpyrazine 97%
Supplier: Apollo Scientific
Warning
OR9783-1G OR9783-5G OR9783-25G
APOSOR9783-1GEA 21.9 EUR
APOSOR9783-1G APOSOR9783-5G APOSOR9783-25G
2-Chloro-6-methylpyrazine 97%
2-Chloro-6-methylpyrazine
Formula: C₅H₅ClN₂
MW: 128.5596 g/mol
Boiling Pt: 83 °C/35mmHg
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00055032
CAS Number: 38557-71-0