Ethyl 2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Warning
OR9789-250MG OR9789-50MG
APOSOR9789-250MGEA 177 EUR
APOSOR9789-250MG APOSOR9789-50MG
Ethyl 2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₆H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00139520
CAS Number: 119448-82-7