2-Acetyl-5-methylthiophene
Supplier: Apollo Scientific
Warning
Synonyms: 1-(5-Methyl-2-thienyl)-1-ethanone
OR9799-10G OR9799-25G OR9799-100G
APOSOR9799-10GEA 21.9 EUR
APOSOR9799-10G APOSOR9799-25G APOSOR9799-100G
2-Acetyl-5-methylthiophene
2-Acetyl-5-methylthiophene
Formula: C₇H₈OS
Boiling Pt: 65…67 °C/1mmHg
Melting Pt: 24…28 °C
Density: 1.119 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014529
CAS Number: 13679-74-8
EINECS: 237-181-2
UN: 3335
ADR: 9,III