4-Chloroaniline
Supplier: Apollo Scientific
Danger
OR9806-1KG OR9806-250G OR9806-5KG
APOSOR9806-1KGEA 168 EUR
APOSOR9806-1KG APOSOR9806-250G APOSOR9806-5KG
4-Chloroaniline
4-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 232 °C
Melting Pt: 67…72 °C
Density: 1.14 g/cm³ (25 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II