Methyl-2-(2-propynyloxy)benzenecarboxylate
Supplier: Apollo Scientific
Warning
OR9820-100MG OR9820-500MG
APOSOR9820-100MGEA 174 EUR
APOSOR9820-100MG APOSOR9820-500MG
Methyl-2-(2-propynyloxy)benzenecarboxylate
Methyl-2-(2-propynyloxy)benzenecarboxylate
Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Boiling Pt: 100 °C/ 0.2mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD06797370
CAS Number: 59155-84-9