3,4-Dihydroxybenzonitrile
Supplier: Apollo Scientific
|
Warning
|
OR9851-25G
OR9851-5G
OR9851-100G
APOSOR9851-25GEA
29
EUR
APOSOR9851-25G
APOSOR9851-5G
APOSOR9851-100G
3,4-Dihydroxybenzonitrile
3,4-Dihydroxybenzonitrile
Formula:
C₇H₅NO₂ MW: 135.12 g/mol Melting Pt: 153…159 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00016436 CAS Number: 17345-61-8 EINECS: 241-367-9 UN: 3439 ADR: 6.1,III |