Ethyl-4-morpholinobenzenecarboxylate
Supplier: Apollo Scientific
Warning
OR9853-1G OR9853-500MG OR9853-5G
APOSOR9853-1GEA 168 EUR
APOSOR9853-1G APOSOR9853-500MG APOSOR9853-5G
Ethyl-4-morpholinobenzenecarboxylate
Ethyl-4-morpholinobenzenecarboxylate
Formula: C₁₃H₁₇NO₃
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD06204958
CAS Number: 19614-15-4