2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole 95%
Supplier: Apollo Scientific
Warning
OR9857-1G OR9857-5G
APOSOR9857-1GEA 106 EUR
APOSOR9857-1G APOSOR9857-5G
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole 95%
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole
Formula: C₉H₇ClN₂O
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00466332
CAS Number: 33575-83-6