3-Nitrosalicylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Nitrosalicylic Acid, 2-Hydroxy-3-nitrobenzoic acid

OR9864-1G OR9864-25G OR9864-100G
APOSOR9864-1GEA 21.9 EUR
APOSOR9864-1G APOSOR9864-25G APOSOR9864-100G
3-Nitrosalicylic acid 98%
3-Nitrosalicylic acid
Formula: C₇H₅NO₅
MW: 183.1183 g/mol
Melting Pt: 142…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00024240
CAS Number: 85-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR