4-Phenylbenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
OR9875-100G
OR9875-25G
OR9875-500G
APOSOR9875-100GEA
43.5
EUR
APOSOR9875-100G
APOSOR9875-25G
APOSOR9875-500G
4-Phenylbenzaldehyde 98%
4-Phenylbenzaldehyde
Formula:
C₁₃H₁₀O Boiling Pt: 184 °C/11mmHg Melting Pt: 57…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006947 CAS Number: 3218-36-8 EINECS: 221-742-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...