5-Bromobenzofuran-2-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR9879-1G
OR9879-500MG
OR9879-5G
APOSOR9879-1GEA
94.4
EUR
APOSOR9879-1G
APOSOR9879-500MG
APOSOR9879-5G
5-Bromobenzofuran-2-carboxylic acid
5-Bromobenzofuran-2-carboxylic acid
Formula:
C₉H₅BrO₃ MW: 241.0400 g/mol Melting Pt: 257…262 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093973 CAS Number: 10242-11-2 UN: 2811 ADR: 6.1 III |