Methyl-2-(4-hydroxy-3-nitrophenyl)acetate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-(4-hydroxy-3-nitrophenyl)acetate
OR9884-1G OR9884-5G
APOSOR9884-1GEA 21.9 EUR
APOSOR9884-1G APOSOR9884-5G
Methyl-2-(4-hydroxy-3-nitrophenyl)acetate 95%
Methyl-2-(4-hydroxy-3-nitrophenyl)acetate
Formula: C₉H₉NO₅
MW: 211.18 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD04125034
CAS Number: 61873-93-6