N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide 95%
Supplier: Apollo Scientific
Warning
Synonyms: Imidazo[1,2-a]pyridine-6-amidoxime
OR9886-1G OR9886-5G
APOSOR9886-1GEA 87 EUR
APOSOR9886-1G APOSOR9886-5G
N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide 95%
N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide
Formula: C₈H₈N₄O
MW: 176.18 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD07366550
CAS Number: 885950-24-3