1-Bromo-2-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromonitrobenzene
OR9891-100G OR9891-10G OR9891-25G
APOSOR9891-100GEA 72.5 EUR
APOSOR9891-100G APOSOR9891-10G APOSOR9891-25G
1-Bromo-2-nitrobenzene 98%
1-Bromo-2-nitrobenzene
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 261 °C
Melting Pt: 40…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007045
CAS Number: 577-19-5
EINECS: 209-409-0
UN: 3459
ADR: 6.1 III