2,2'-Dibromoacetophenone 90%
Supplier: Apollo Scientific
Danger
OR9900-1G OR9900-25G OR9900-5G
APOSOR9900-1GEA 43.5 EUR
APOSOR9900-1G APOSOR9900-25G APOSOR9900-5G
2,2'-Dibromoacetophenone 90%
2,2'-Dibromoacetophenone
Formula: C₈H₆Br₂O
Boiling Pt: 90…92 °C/0.02mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00044660
CAS Number: 49851-55-0
UN: 3265
ADR: 8,II