4-(2-Bromoethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
OR9912-1G OR9912-500MG OR9912-5G
APOSOR9912-1GEA 198 EUR
APOSOR9912-1G APOSOR9912-500MG APOSOR9912-5G
4-(2-Bromoethoxy)benzaldehyde 95%
4-(2-Bromoethoxy)benzaldehyde
Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00021074
CAS Number: 52191-15-8