Ethyl-2,4-dimethylpyrrole-3-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate
OR9926-1G OR9926-5G
APOSOR9926-1GEA 21.9 EUR
APOSOR9926-1G APOSOR9926-5G
Ethyl-2,4-dimethylpyrrole-3-carboxylate 98%
Ethyl 2,4-dimethylpyrrole-3-carboxylate
Formula: C₉H₁₃NO₂
MW: 167.2050 g/mol
Melting Pt: 122…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01365811
CAS Number: 2199-51-1