2-Methoxy-6-methylpyridine 98%
Supplier: Apollo Scientific
Warning
OR9940-100G OR9940-25G
APOSOR9940-100GEA 206 EUR
APOSOR9940-100G APOSOR9940-25G
2-Methoxy-6-methylpyridine 98%
2-Methoxy-6-methylpyridine
Formula: C₇H₉NO
MW: 123.16 g/mol
Boiling Pt: 156 °C
Density: 1.01 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD04112507
CAS Number: 63071-03-4
UN: 1993
ADR: 3 III