3-Methylanthranil acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-3-methylbenzoic acid
OR9942-100G OR9942-25G
APOSOR9942-100GEA 30.6 EUR
APOSOR9942-100G APOSOR9942-25G
3-Methylanthranil acid 98%
3-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 315
Melting Pt: 174…177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007745
CAS Number: 4389-45-1
EINECS: 224-505-2